
4-Quinolinecarbaldehyde 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4363-93-3
:
Formula:C₁₀H₇NO
MDL Number:MFCD00006781
:157.17 g/mol
Boiling Pt:122 - 123 °C (4 -rr)
Melting Pt:45…52 °C
Flash Pt:>112 °C
Storage Temperature:Refrigerator
EINECS:224-453-0
:GHS07
Specification Test Results
- Appearance:Off-white to yellow to light brown Crystalline powder and/or chunks or solid
- Infrared spectrum:Conforms
- Melting point:45 to 52 °C
- GC:≥94 %