Order Entry
Finland
ContactUsLinkComponent
(1S)-(-)-α-Pinene for synthesis, Sigma-Aldrich®
(1S)-(-)-α-Pinene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
(1S)-(-)-α-Pinene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    -62 °C
  • Molar Mass:
    136.23 g/mol
  • Boiling Point:
    155 - 156 °C
  • Vapor Pressure:
    5 hPa (25 °C)
  • Flash Point:
    32 °C
  • Refractive Index:
    1.446 (20 °C, 589 nm)
  • Explosion Limit:
    0.8 %(V)
  • Density:
    0.86 g/cm3 (20 °C)
  • Ignition Point:
    255 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

Related Information