(1S)-(-)-α-Pinene 98%
:7785-26-4
:
:C₁₀H₁₆
:MFCD00064145
:136.24 g/mol
:12,07599
:155…156 °C (760 mmHg)
:–64 °C
:0.8595 g/cm³
:33 °C
:232-077-3
:2368
:3,III
:GHS07
:GHS08
:GHS09
:GHS02
:Ambient
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4640 to 1.4680 (20°C, 589 nm)
- GC:≥97.5 %
- Color scale:≤30 APHA
- Specific gravity:(20°C) 0.858 to 0.861
- Specific optical rotation:-35° to -45° (20°C, 589 nm) (Neat)