(1R)-(+)-α-Pinene 98%, 80% ee
:7785-70-8
:
:C₁₀H₁₆
:MFCD00001346
:136.24 g/mol
:12,07599
:155 °C (1013 hPa)
:–62 °C
:0.86 g/cm³
:33 °C
:232-087-8
:2368
:3,III
:GHS07
:GHS08
:GHS09
:GHS02
:Refrigerator
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4650 to 1.4670 (20°C, 589 nm)
- GC:≥97.5 %
- Enantiomeric excess:≥75 %
- Specific gravity:(20°C) 0.855 to 0.865
- Specific optical rotation:+40° to +47° (20°C, 589 nm) (neat)