Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Urea
Urea 40% in aqueous solution
Urea 40% in aqueous solution
SCAR6083100
:
Scharlau
:
Urea 40% in aqueous solution
SCAR6083100
:
Scharlau
:
Pack type:
Glass bottle
Pk:
100 ml
Seq:
2440
:
:
60.06 g/mol
:
Ambient
:
200-315-5
:
57-13-6
:
NH₂CONH₂
:
MFCD00008022