Order Entry
Finland
ContactUsLinkComponent
 
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture 99+%
Catalog # ACRO406040050
CAS Number:  
undefined
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture 99+%
Catalog # ACRO406040050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    164 to 168 °C
  • GC:
    ≥99.0 %

Specification Test Results

About this item

 :
 :172.18 g/mol
Melting Pt:165…168 °C
Storage Temperature:Ambient
EINECS:214-068-6
CAS Number:1076-97-7
Formula:C₈H₁₂O₄
MDL Number:MFCD00001465