Trityl chloride
Catalog # MOLE22303758-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:278.78 g/mol
Boiling Pt:374 °C (1013 hPa)
Melting Pt:109…112 °C
Density:1.25 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-986-4
UN:3261
ADR:8,II
:GHS05
CAS Number:76-83-5
Formula:C₁₉H₁₅Cl
MDL Number:MFCD00000813