Order Entry
Finland
ContactUsLinkComponent
Cumene for synthesis, Sigma-Aldrich®
Cumene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Cumene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.05 g/l (20 °C)
  • Melting Point:
    -96 °C
  • Molar Mass:
    120.19 g/mol
  • Boiling Point:
    153 °C (1013 hPa)
  • Vapor Pressure:
    5.3 hPa (20 °C)
  • Flash Point:
    0.8 - 6.0 %(V)
  • Refractive Index:
    31 °C
  • Density:
    0.86 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable
  • Ignition Point:
    420 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

Related Information