Cumene 99%
:98-82-8
:
:C₉H₁₂
:MFCD00008881
:13,02644
:151 °C (1013 hPa)
:–96 °C
:0.862 g/cm³
:31 °C
:1918
:3,III
:120.19 g/mol
:202-704-5
:GHS02
:GHS07
:GHS08
:GHS09
:Ambient
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4900 to 1.4930 (20°C, 589 nm)
- GC:≥98.5 %
- HPLC:≥97.5 %