

α,α,α-Trifluoro-3-nitrotoluene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-46-4
:
Formula:C₇H₄F₃NO₂
MDL Number:MFCD00007260
:191.11 g/mol
Boiling Pt:201…205 °C
Melting Pt:–5 °C
Density:1.435
Flash Pt:87 °C (188 °F)
Storage Temperature:Ambient
EINECS:202-670-1
UN:2306
ADR:6.1,II
:GHS06
:GHS07