
4-Chloro-3-nitrobenzotrifluoride 98%
ACRO109701000
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4880 to 1.4910 (20°C, 589 nm)
- GC:≥97.5 %
:
:225.55 g/mol
:223 °C (1013 hPa)
:–2 °C
:1.511 g/cm³
:101 °C
:Ambient
:204-451-6
:2307
:6.1,II
:GHS07
:121-17-5
:C₇H₃ClF₃NO₂
:MFCD00007084