
m-Toluic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99-04-7
:
Formula:C₈H₈O₂
MDL Number:MFCD00002523
:136.15 g/mol
Merck Index:14,9535
Boiling Pt:263 °C
Melting Pt:108…110 °C
Density:1.050
Flash Pt:150 °C (302 °F)
EINECS:202-723-9
:GHS07
Storage Temperature:Ambient