Order Entry
Finland
Orders LinkContactUsLinkComponent
α-Bromo-3-nitrotoluene

α-Bromo-3-nitrotoluene

Glass bottle for solids, 5 g
Catalog # MOLEM21918054
Marketsource
Supplier:   Molekula
α-Bromo-3-nitrotoluene
Catalog # MOLEM21918054
Supplier:  Molekula
Marketsource
CAS Number:  3958-57-4
α-Bromo-3-nitrotoluene

Glass bottle for solids, 5 g

CAS Number:  3958-57-4
Price
75,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  α-Bromo-m-nitrotoluene, 1-(Bromomethyl)-3-nitrobenzene, 3-Nitrobenzyl bromide, m-Nitrobenzyl bromide
  • CAS Number:3958-57-4
  • Formula:C₇H₆BrNO₂
  • MDL Number:MFCD00007271
  • Molecular Weight:216.03 g/mol
  • Boiling Pt:115…118 °C (0.8 mmHg)
  • Melting Pt:59…60 °C
  • EINECS:223-557-3
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.