Ämpärit
Supplier: BOCHEM
Jump to:
About this item
Buckets, stainless steel 18/10.
- Very good chemical resistance, very good thermal resistance
- High quality, very stable
- Matching lids available optionally
For use in laboratories and industry. Graduated with handle. Seamless handle tabs and smooth surfaces for fast, easy cleaning.
11 Options Available Below
Product Details & Documents
Buckets, stainless steel 18/10.
- Very good chemical resistance, very good thermal resistance
- High quality, very stable
- Matching lids available optionally
For use in laboratories and industry. Graduated with handle. Seamless handle tabs and smooth surfaces for fast, easy cleaning.
Product Family Options
Ämpäri pohjareunuksella
Product Information
Ämpäri ilman pohjareunusta
Product Information
- KuvausKapasiteettiØ Ulk.KorkeusAvailabilityPrice
Lids for buckets
Product Information
- KuvausKapasiteettiØ Ulk.KorkeusAvailabilityPrice
- Kansi mitta-asteikollisiin ämpäreihin 10 l, ruostumatonta terästä 18/10, Ø 280 mmLog in for availability1 * 1 Items58,70 €
- Kansi mitta-asteikollisiin ämpäreihin 12 l, ruostumatonta terästä 18/10, Ø 300 mmLog in for availability1 * 1 Items62,50 €
- Kansi mitta-asteikollisiin ämpäreihin 15 l, ruostumatonta terästä 18/10, Ø 310 mmLog in for availability1 * 1 Items68,00 €