Order Entry
Finland
Orders LinkContactUsLinkComponent
Brushes for flasks
Brushes for flasks
Supplier:   BOCHEM
Brushes for flasks
Supplier:  BOCHEM
Certificates
Brushes for flasks
Brushes for flasks
4 Options Available Below
Product Details & Documents
Brushes for flasks, iron wire, zinced with natural bristles.
Product Family Options
Product Information
  • Bristle colour
    Brush head length
    Brush head Ø
    Handle material
    Total length
    Handle length
    Availability
    Price
  • Natural
    130 mm
    50 mm
    Wire
    380 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6,90 €
    6,90 €
    1 * 1 Items
       6,90 €
  • Natural
    140 mm
    55 mm
    Wire
    420 mm
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7,60 €
    7,60 €
    1 * 1 Items
       7,60 €
  • Natural
    120 mm
    63 mm
    Wire
    480 mm
    360 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9,50 €
    9,50 €
    1 * 1 Items
       9,50 €
  • Natural
    190 mm
    80 mm
    Wire
    550 mm
    360 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 14,00 €
    14,00 €
    1 * 1 Items
       14,00 €