


3-Penten-2-one tech. 85%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:625-33-2
:
Formula:C₅H₈O
MDL Number:MFCD00009290
:84.12 g/mol
Boiling Pt:119…121 °C
Density:0.862
Flash Pt:21 °C (69 °F)
EINECS:210-888-3
UN:1992
ADR:3,II
:GHS02
:GHS06
:GHS07
Storage Temperature:Ambient