
4-Hydroxyphenylboronic acid pinacol ester 97%
Catalog # ACRO366420050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Yellow to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:112 to 117 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:220.08 g/mol
Melting Pt:112…117 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:269409-70-3
Formula:C₁₂H₁₇BO₃
MDL Number:MFCD02093756