Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Aminophenylboronic acid pinacol ester
4-Aminophenylboronic acid pinacol ester 98%
4-Aminophenylboronic acid pinacol ester 98%
4-Aminophenylboronic acid pinacol ester 98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
4-Aminophenylboronic acid pinacol ester 98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
214360-73-3
:
Formula:
C₁₂H₁₈BNO₂
MDL Number:
MFCD02093721
:
219.09 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH51666
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation