Erythrosin B disodium salt, pure, certified
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16423-68-0
:568-63-8
Formula:C₂₀H₆I₄Na₂O₅
MDL Number:MFCD00144257
:879.86 g/mol
Melting Pt:>300 °C
EINECS:240-474-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Red to brown Powder
- Infrared spectrum:Conforms
- Dye content:≥80 %
- UV Lambda max:524 to 527 nm
- Absorption Ratio P-15/P+15:1.15 to 1.55
- BSC Certification:Passes