1-Acetyl-4-piperidone 97%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: N-Acetyl-4-piperidone
- CAS Number:32161-06-1
- Formula:C₇H₁₁NO₂
- MDL Number:MFCD00006190
- Molecular Weight:141.17 g/mol
- Density:1.14 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:250-937-6
2 Options Available Below
Product Details & Documents
Synonyms: N-Acetyl-4-piperidone
- CAS Number:32161-06-1
- Formula:C₇H₁₁NO₂
- MDL Number:MFCD00006190
- Molecular Weight:141.17 g/mol
- Density:1.14 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:250-937-6
Documents
ACRO16682
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear orange Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.5015 to 1.5035 (20°C, 589 nm)
- GC:≥96.0 %
- Clarity of solution:Passes test
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



