Brilliant cresyl blue, dark green powder C.I. 51010
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:81029-05-2
:10127-36-3
Formula:C₃₄H₄₂Cl₄N₈O₂Zn
MDL Number:MFCD00148901
:801.96 g/mol
EINECS:225-203-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test :Passes
- UV/Vis: l max (ethanol):For Information Only