Lead(II) acetate
Glass bottle for solids, 5 g
Catalog # FFCHFF61720005
Supplier: FF CHEMICALS
Jump to:
Product Details & Documents
Synonyms: Acetic acid Lead(II) salt
- CAS Number:301-04-2
- Formula:(H₃CCOO)₂Pb
- MDL Number:MFCD00012452
- Molecular Weight:325.29 g/mol
- Merck Index:12,05411
- Melting Pt:280 °C
- Density:2.55 g/cm³ (20 °C)
- EINECS:206-104-4
- UN:1616
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



