Order Entry
Finland
ContactUsLinkComponent
 
 
 
4-Nitroaniline for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
4-Nitroaniline for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Hill Formula:
    C₆H₆N₂O₂
  • Chemical formula:
    H₂NC₆H₄NO₂
  • Molar Mass:
    138.13 g/mol
  • Boiling point:
    331 °C (1013 hPa) (decomposition)
  • Density:
    1.44 g/cm³ (20 °C)
  • Flash point:
    199 °C
  • Ignition temperature:
    500 °C
  • Melting Point:
    148 °C
  • pH value:
    (0.5 g/l, H₂O, 20 °C) neutral
  • Vapor pressure:
    1.33 hPa (142 °C)
  • Bulk density:
    630 kg/m³
  • Solubility:
    0.5 g/l (20 °C)
  • Storage class:
    6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
  • WGK:
    WGK 3 highly water endangering
  • R Phrase:
    R 23/24/25-33-52/53
  • S Phrase:
    S 28-36/37-45-61