Burettes, straight stopcock, PYREX®
Supplier: SciLabware
Certificates
About this item
Borosilicate glass with interchangeable PTFE key, Class B.
- Calibrated to deliver ‘Ex’
- PTFE key for grease free performance
- White enamel graduations for good visibility with dark coloured solutions
Certifications: ISO 385