Magnetic stirring discs
Supplier: KARTELL
Certificates
About this item
PTFE coated magnetic stirring bars with convex shape for good centring action.
- Effective stirring action
- Suitable for food stuff
PTFE coated magnetic stirring bars with convex shape for good centring action.