Order Entry
Finland
Orders LinkContactUsLinkComponent
Bottle Carriers
Supplier:   BURKLE
Bottle Carriers
Supplier:  BURKLE
Certificates
Bottle Carriers
2 Options Available Below
Product Details & Documents
Made of PP or HDPE.
  • Bottle transport container (PP) for up to 6 bottles, bottle transport box for up to 8 bottles
  • Bottle transport box (HDPE): suitable for food; With impermeable base suitable as a collecting basin
Product Family Options
Carrier box
Product Information
  • Colour
    For
    Compartment size
    Container max. Ø
    Availability
    Price
  • BURK0361-0008
    Bottle carrier, HDPE, for food with closed floor
    Bottle carrier, HDPE, for food with closed floor
    Grey
    Up to eight bottles
    390×110 mm
    95 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 48,90 €
    48,90 €
    1 * 1 Items
       48,90 €
Product Information
  • Colour
    For
    Compartment size
    Container max. Ø
    Availability
    Price
  • BURK0361-0006
    Bottle carrier, HDPE
    Bottle carrier, HDPE
    Grey
    Up to six bottles
    95 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 22,80 €
    22,80 €
    1 * 1 Items
       22,80 €