Order Entry
Finland
Orders LinkContactUsLinkComponent
2,5-Dichlorophenol ≥98%
2,5-Dichlorophenol ≥98%
2,5-Dichlorophenol ≥98%
2,5-Dichlorophenol ≥98%
Purity: 98% CAS No.: 583-78-8 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odour: Phenol-like Melting Point: 55-58° Boiling Point: 211° Beilstein Reference: 1907692 UN No.: 2020 MDL No.: MFCD00002174
  •  :583-78-8
  •  :C₆H₄Cl₂O
  •  :MFCD00002174
  •  :163 g/mol
  •  :211 °C
  •  :55…58 °C
  •  :Ambient
  •  :209-520-4
  •  :2020
  •  :6.1,III
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    25 g
    1 * 25 g
    1 * 25 g - 50,40 €
    50,40 €
    1 * 25 g
       50,40 €
  • Glass bottle for solids
    100 g
    1 * 100 g
    1 * 100 g - 125,00 €
    125,00 €
    1 * 100 g
       125,00 €
  • Glass bottle for solids
    500 g
    1 * 500 g
    1 * 500 g - 420,00 €
    420,00 €
    1 * 500 g
       420,00 €