3,4-Dichlorophenol ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 95-77-2 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odour: Phenol-like Melting Point: 64-68° Boiling Point: 252-255° Beilstein Reference: 1907693 UN No.: 2020 MDL No.: MFCD00002258
- CAS Number:95-77-2
- Formula:C₆H₄Cl₂O
- MDL Number:MFCD00002258
- Molecular Weight:163 g/mol
- Boiling Pt:252…255 °C
- Melting Pt:64…68 °C
- Density:1.6 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:202-450-5
- UN:2020
- ADR:6.1,III
- :Density: 1.6 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 95-77-2 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odour: Phenol-like Melting Point: 64-68° Boiling Point: 252-255° Beilstein Reference: 1907693 UN No.: 2020 MDL No.: MFCD00002258
- CAS Number:95-77-2
- Formula:C₆H₄Cl₂O
- MDL Number:MFCD00002258
- Molecular Weight:163 g/mol
- Boiling Pt:252…255 °C
- Melting Pt:64…68 °C
- Density:1.6 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:202-450-5
- UN:2020
- ADR:6.1,III
- :Density: 1.6 g/cm³ (20 °C)



