4-Chloroanisole ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:623-12-1
:
Formula:C₇H₇ClO
MDL Number:MFCD00000616
:142.58 g/mol
Boiling Pt:198…200 °C
Melting Pt:–18 °C
Density:1.170
Flash Pt:78 °C (172 °F)
Storage Temperature:Ambient
EINECS:210-772-2