Order Entry
Finland
ContactUsLinkComponent
1,2-Dichloro-3-nitrobenzene for synthesis, Sigma-Aldrich®
Irritant
Environment
1,2-Dichloro-3-nitrobenzene for synthesis, Sigma-Aldrich®
Catalog # 8.40100.0250
Supplier:  Merck
CAS Number:  
Irritant
Environment
1,2-Dichloro-3-nitrobenzene for synthesis, Sigma-Aldrich®
Catalog # 8.40100.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    0.067 g/l (20 °C)
  • Melting Point:
    61 °C
  • Molar Mass:
    192 g/mol
  • Boiling Point:
    270 °C (1013 hPa)
  • Vapor Pressure:
     - 1.3 hPa (50 °C)
  • Flash Point:
    152 °C
  • Density:
    1.45 g/cm3 (80 °C)

Specification Test Results

About this item

 :
 :192 g/mol
Boiling Pt:257 °C (1013 hPa)
Melting Pt:61 °C
Density:1.721 g/cm³ (14 °C)
Storage Temperature:Ambient
EINECS:221-717-7
UN:1578
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
 :GHS07
 :GHS09
CAS Number:3209-22-1
Formula:C₆H₃Cl₂NO₂
MDL Number:MFCD00007062