Eyewash station for tap, EyesFresh
Catalog # 118-0275
Supplier: BURKLE
Specifications
- Description:EyesFresh mini eyewash station, tap attachment, for normal use according to DIN 12899
- Pk:1
Specifications
About this item
This mini eyewash station is ideal for laboratories, filling stations or all work places where aggressive substances are used.
- Flexible usage as it can be attached to most water taps
- No refilling needed as the water from the tap is used
It can be easily attached to any standard cold water tap. For normal use of the tap, the user just needs to press the button and the function changes back from the eye wash (returns automatically to eyewash-function when the tap is closed).
Can be fitted onto taps with M22×1, 1/2" or 3/8" outer thread.
Certifications: With sufficient water pressure provided, performance according to DIN EN 15154-2.