
4-Aminosalicylic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65-49-6
:
Formula:C₇H₇NO₃
MDL Number:MFCD00007789
:153.14 g/mol
Merck Index:13,00476
Boiling Pt:347 °C (1013 hPa)
Melting Pt:147 °C (dec.)
Density:1,545 g/cm³ (20 °C)
EINECS:200-613-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Beige Powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥98.5 %
- GC:≥96.0 %