Order Entry
Finland
ContactUsLinkComponent
3-Amino-4-hydroxybenzoic acid
Irritant
3-Amino-4-hydroxybenzoic acid
Catalog # MOLEM58031924
Supplier:  Molekula
CAS Number:  
Irritant
3-Amino-4-hydroxybenzoic acid
Catalog # MOLEM58031924
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :153.14 g/mol
Boiling Pt:347 °C (1013 hPa)
Melting Pt:201 °C
EINECS:216-390-2
 :GHS07
Storage Temperature:Ambient
CAS Number:1571-72-8
Formula:C₇H₇NO₃
MDL Number:MFCD00007697
Boiling Pt:347 °C (1013 hPa)