Order Entry
Finland
ContactUsLinkComponent
Gas washing bottles, DURAN®
Gas washing bottles, DURAN®
Catalog # SCOT257040101
Supplier:  DWK Life Sciences
undefined
Gas washing bottles, DURAN®
Catalog # SCOT257040101
Supplier:  DWK Life Sciences
Specifications
  • Thread:
    GL 45
  • Capacity:
    500 ml
  • Pk:
    1
Specifications
About this item
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw cap system with Drechsel-head with filter disc, adjustable immersion.