Order Entry
Finland
Orders LinkContactUsLinkComponent
Ethyl 4-hydroxybenzoate 99%

Ethyl 4-hydroxybenzoate 99%

Ethyl 4-hydroxybenzoate 99%
Certificates
Ethyl 4-hydroxybenzoate  99%

4 Options Available Below

Product Details & Documents
Synonyms:  Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
  • CAS Number:120-47-8
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002353
  • Molecular Weight:166.18 g/mol
  • Merck Index:13,03869
  • Boiling Pt:297…298 °C (760 mmHg)
  • Melting Pt:115…118 °C
  • Density:1,078 g/cm³ (131 °C)
  • Storage Temperature:Ambient
  • EINECS:204-399-4

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light cream Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    115 to 118 °C
  • GC:
    ≥98.5 %
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.