Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Nitrobenzonitrile
4-Nitrobenzonitrile 97%
4-Nitrobenzonitrile 97%
:
Thermo Fisher Scientific
4-Nitrobenzonitrile 97%
:
Thermo Fisher Scientific
:
619-72-7
:
:
C₇H₄N₂O₂
:
MFCD00007279
:
148.12 g/mol
:
144…149 °C
:
Ambient
:
210-610-0
:
3439
:
6.1,II
:
GHS06
Appearance:
Yellow Crystalline powder
Infrared spectrum:
Conforms
Melting point:
144 to 149 °C
GC:
≥96.0 %
ACRO12848