2,5-Dihydroxybenzaldehyde
Glass bottle for solids, 5 g
Catalog # MOLE31698533-5G
Marketsource
Supplier: Molekula
2,5-Dihydroxybenzaldehyde
Catalog # MOLE31698533-5G
Supplier: Molekula
Marketsource
CAS Number: 1194-98-5
Jump to:
Product Details & Documents
- CAS Number:1194-98-5
- Formula:C₇H₆O₃
- MDL Number:MFCD00003333
- Molecular Weight:138.12 g/mol
- Boiling Pt:60…65 °C (12 mmHg)
- Melting Pt:99 °C
- Density:0,859 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:214-789-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



