4-Hydroxybenzaldehyde
Glass bottle for solids, 100 mg
Catalog # PROOMM0460.14
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:123-08-0
- Formula:C₇H₆O₂
- MDL Number:MFCD00006939
- Molecular Weight:122.12 g/mol
- Merck Index:13,04836
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:116 °C
- Density:1.13 g/cm³ (20 °C)
- Flash Pt:174 °C
- Storage Temperature:Ambient
- EINECS:204-599-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



