
Vanillin, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:121-33-5
:
Formula:C₈H₈O₃
MDL Number:MFCD00006942
:152.15 g/mol
Merck Index:13,09998
Boiling Pt:285 °C (1013 hPa)
Melting Pt:∼82 °C
Density:1.056 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:153 °C
EINECS:204-465-2
:GHS07
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberMERP1.08510.1000_TDS_EN ProductPage_108510
Product specification
msds_108510
SDS
Frequently Bought Together