Burettes, straight stopcock, class B, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Straight stopcock
- With glass key
- Marks and inscription in blue
- Calibration is based on the poured out volume ('Ex')
Class B limit is roughly one and a half times wider than for class AS. The tolerances are thus more strict than specified by DIN. Typical application: titrations.
Certifications: Complies to the DIN ISO 385.