2-Nitrobenzaldehyde
Glass bottle for solids, 100 mg
Catalog # PROOMM0003.03
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:552-89-6
- Formula:C₇H₅NO₃
- MDL Number:MFCD00007132
- Molecular Weight:151.12 g/mol
- Merck Index:12,06684
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:41…43 °C
- Density:1.35 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:209-025-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



