Order Entry
Finland
Orders LinkContactUsLinkComponent
2-Nitrobenzaldehyde

2-Nitrobenzaldehyde

Glass bottle for solids, 100 g
Catalog # MOLE10585537-100G
Marketsource
Supplier:   Molekula
2-Nitrobenzaldehyde
Catalog # MOLE10585537-100G
Supplier:  Molekula
Marketsource
CAS Number:  552-89-6
2-Nitrobenzaldehyde

Glass bottle for solids, 100 g

CAS Number:  552-89-6
Price
76,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:552-89-6
  • Formula:C₇H₅NO₃
  • MDL Number:MFCD00007132
  • Molecular Weight:151.12 g/mol
  • Merck Index:12,06684
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:41…43 °C
  • Density:1.35 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:209-025-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.