Order Entry
Finland
Orders LinkContactUsLinkComponent
2-Nitrobenzaldehyde ≥98%

2-Nitrobenzaldehyde ≥98%

2-Nitrobenzaldehyde ≥98%
Certificates
2-Nitrobenzaldehyde ≥98%
2-Nitrobenzaldehyde ≥98%

3 vaihtoehtoa saatavilla alla

Product Details & Documents

Purity: 98+% CAS No.: 552-89-6 Molecular Formula: C7H5NO3 Formula Weight: 151.12 Flash Point: 110°(230°F) Melting Point: 43-46° Boiling Point: 153°/23mm Beilstein Reference: 742624 Merck Reference: 14,6587 MDL No.: MFCD00007132

  • CAS numero:552-89-6
  • Kaava:C₇H₅NO₃
  • MDL-numero:MFCD00007132
  • Molecular Weight:151.12 g/mol
  • Merck-indeksi:14,6587
  • Kiehumispiste:153 °C (23 mmHg)
  • Sulamispiste:43…46 °C
  • Tiheys:1.35 g/cm³ (25 °C)
  • Leimahduspiste:>110 °C (230 °F)
  • Varastointilämpötila:Ambient
  • EINECS:209-025-3
Product Family Options

Tuotetiedot

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.