Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bottles
Weighing Bottles
Weighing bottles, glass, DURAN®
Weighing bottles, glass, DURAN®
:
DWK Life Sciences
Weighing bottles, glass, DURAN®
:
DWK Life Sciences
DURAN® Borosilicate glass 3.3, clear.
Interchangeable lids with ground joint
Writing area in white
Available in low or tall form