
1-Bromo-4-(trifluoromethoxy)benzene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:407-14-7
:
Formula:C₇H₄BrF₃O
MDL Number:MFCD00040834
:241.01 g/mol
Boiling Pt:153 °C (1013 hPa)
Density:1.62 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-979-2
UN:3334
ADR:9,
:M-Clarity™ quality level = MQ200
:GHS07
Specification Test Results
- Solubility:11.7 mg/l (20 °C)
- Molar Mass:241.01 g/mol
- Boiling Point:91 - 92 °C (100 hPa)
- Vapor Pressure:20 hPa (55 °C)
- Flash Point:67 °C
- Density:1.62 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together