Toluenediisocyanate mixture of isomers ≥98.0% (by GC) (2,4- ∼ 80%, 2,6- ∼ 20%)
Supplier: TCI
Certificates
About this item
CAS Number:26471-62-5
:584-84-9
Formula:C₉H₆N₂O₂
MDL Number:MFCD01778251
Boiling Pt:251°C
Melting Pt:14°C
Flash Pt:137°C
Storage Temperature:Refrigerator
EINECS:247-722-4
UN:2078
ADR:6.1,II
:174.16
:GHS05
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 % (total of isomer)