

1-Bromo-2-fluorobenzene 99%
:1072-85-1
:
:C₆H₄BrF
:MFCD00000282
:175 g/mol
:155 °C (1013 hPa)
:1.6 g/cm³
:43 °C
:Ambient
:214-018-3
:1993
:3,III
:GHS07
:GHS02
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5327 to 1.5347 (20°C, 589 nm)
- GC:≥98.5 %
- Titration with NaOH:≥98.5 %