
2,5-Dibromo-1-fluorobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1435-52-5
:
Formula:C₆H₃Br₂F
MDL Number:MFCD00010603
Boiling Pt:216 °C
Melting Pt:33…35 °C
Flash Pt:101 °C (213 °F)
:253.9 g/mol
:GHS07
Storage Temperature:Ambient