Order Entry
Finland
ContactUsLinkComponent
 
Irritant
2-Bromo-5-nitrobenzoic acid
Catalog # ACRO342290050
CAS Number:  
undefined
Irritant
2-Bromo-5-nitrobenzoic acid
Catalog # ACRO342290050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light beige to light brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    178 to 184 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :246.02 g/mol
Boiling Pt:371 °C (1013 hPa)
Melting Pt:180…181 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:943-14-6
Formula:C₇H₄BrNO₄
MDL Number:MFCD00134558