1-Hexyne 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:693-02-7
:
Formula:C₆H₁₀
MDL Number:MFCD00009504
:82.15 g/mol
Boiling Pt:71…72 °C (760 mmHg)
Melting Pt:–132 °C
Density:0.71 g/cm³
Flash Pt:–021 °C
Storage Temperature:Refrigerator
EINECS:211-736-9
UN:3295
ADR:3,II
:GHS07
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3980 to 1.4000 (20°C, 589 nm)
- GC:≥97.5 %